missing translation for 'onlineSavingsMsg'
Learn More

Agarose LE, Molecular Biology Grade, Ultrapure, Thermo Scientific Chemicals

Product Code. 15825208
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Granular Free-flowing Powder
Quantity 25 g
Packaging Plastic Bottle
Grade Molecular Biology
Chemical Name or Material Agarose LE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.