missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™

Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max.

94.65 € - 397.00 €

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10766834
View Documents
Fisher BioReagents
BP160-100
100 g Poly Bottle
94.65 €
100g
In Stock
to see stock.
Edge
Add to basket
 
10366603
View Documents
Fisher BioReagents
BP160-500
500 g Poly Bottle
397.00 €
500g
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Agarose, C12H18O9, CAS Number-9012-36-6, 500g, Gray, Cream, DNase free, 0.09 to 0.13, Pass Test, 34.5 deg. to 37.5 deg.C, 1200g/cm2min., Molecular Biology, Low-EEO/Multi-Purpose, Poly Bottle, Powder/Solid, DNase- and RNase-Free, Room temperature, 0.15% max.

Specifications

Chemical Identifiers

9012-36-6
C12H18O9

Specifications

Powder/Solid
Cream
0.09 to 0.13
1200g/cm2min.
DNase- and RNase-Free
9012-36-6
DNase free
34.5° to 37.5°C
C12H18O9
Agarose, Low-EEO/Multi-Purpose
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room temperature

SDS
Documents

Documents

Product Certifications