missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose ME, for electrophoresis of macromolecules

Product Code. 11482688
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agarose is frequently used in molecular biology for the separation of large molecules, especially DNA, by electrophoresis. Slabs of agarose gels (usually 0.7 - 2%) for electrophoresis are readily prepared by pouring the warm, liquid solution into a mold. A wide range of different agaroses, of varying molecular weights and properties are commercially available for this purpose. Used in high resolution protein electrophoresis, immunoelectrophoresis, electro immunoassays such as rocket assays, counter current electrophoresis.

Solubility
Soluble in water (10 g/L at 80°C).

Notes
Gel point 36°. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Air, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Quantity 25 g
Solubility Information Soluble in water (10g/L at 80°C).
Chemical Name or Material Agarose ME, for electrophoresis of macromolecules

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'