missing translation for 'onlineSavingsMsg'
Learn More
Agarose, Molecular biology reagent grade, MP Biomedicals™ Product Code.: 11476401
Estimated Shipment: 27-02-2025
to see stock.
Add to basket

Agarose, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11476401
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11476401

Brand: MP Biomedicals 0219398380

Estimated Shipment: 27-02-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 9012-36-6
Synonym 3, 6-Anhydro-a-L-galacto-β-D-galactan, Sepharose

Specifications

Physical Form Powder
Color White
Quantity 100 g
Grade Molecular Biology
Chemical Name or Material Agarose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Agarose, Molecular biology reagent grade, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.