missing translation for 'onlineSavingsMsg'
Learn More

Agarose, Molecular biology reagent grade, MP Biomedicals™

Product Code. 11496401
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 9012-36-6
Synonym 3, 6-Anhydro-a-L-galacto-β-D-galactan, Sepharose

Specifications

Physical Form Powder
Color White
Quantity 500 g
Grade Molecular Biology
Chemical Name or Material Agarose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.