missing translation for 'onlineSavingsMsg'
Learn More

Agarose, MP Biomedicals™

Product Code. 11491552
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Agarose Genetic Technology Grade, White Powder

  • Agarose is used to separate nucleic acids electrophoretically because agarose gels have larger pore sizes than cross linked acrylamide gels at low concentrations.
  • Agarose gels tend to be more solid, but less elastic, than cross-linked acrylamide gels.
  • Agarose gels are also used in immunoelectrophoresis (IEP) and double-diffusion Ouchterlony plates to demonstrate antibody-antigen reactions.
  • Agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C24H38O19
Molecular Weight (g/mol) 630.55
MDL Number MFCD00081294
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N
Synonym 3,6-Anhydro-a-L-galacto-b-D-galactan, Sepharose

Specifications

Physical Form Powder
Melting Point 87.3°C to 89.5°C
Color White
pH 5.5 to 7.5 (1.5% aq. soln.)
Quantity 100 g
Electroendosmosis (EEO) 0.11 - 0.17
Gelation Temperature 35 - 37°C
Gel Strength > 2000gm/cm2(1.5%gel)
Solubility Information 1g in 100 mL, in water.
Grade Genetic Technology
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'