missing translation for 'onlineSavingsMsg'
Learn More

Agarose Multipurpose, MP Biomedicals™

Product Code. 11436960
Click to view available options
Quantity:
3 x 100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Agarose Multipurpose, Molecular Biology Grade, White powder

  • Agarose is a natural polysaccharide isolated from agar or agar-bearing marine algae.
  • Its physical and chemical properties are particularly suited for use as a gel medium for diffusion and electrokinetic movement of biomolecules.
  • Agarose is essentially biologically inert and forms relatively clear, transparent gels.
  • Agarose based gels offer several advantages: High gel strength permits use of low concentrations.
  • The macroporous nature of the gels allows rapid diffusion of high molecular weight molecules. Agarose is Non-toxic.
  • Agarose gels are thermally reversible facilitating sample recovery.
  • Staining and destaining may be done rapidly with minimal background.
  • Because agarose forms macroporous gels that are electrically nonionic, it is an excellent medium for Isoelectric Focusing (IEF).
  • A selection of different formulated agaroses with specific qualities for unique applications are available from MP.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C24H38O19
Molecular Weight (g/mol) 630.55
MDL Number MFCD00081294
InChI Key MJQHZNBUODTQTK-WKGBVCLCSA-N

Specifications

Physical Form Powder
Color White
Quantity 3 x 100 g
Synonym Galactan Hydrocolloid, 3,6-Anhydro-α-L-galacto-β-D-galactan
Solubility Information Soluble in water (1.5g plus 100 mL)
Grade Molecular Biology
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.