missing translation for 'onlineSavingsMsg'
Learn More

Agarose (Protein Electrophoresis Grade), Fisher BioReagents™

Product Code. 10775684
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Agarose (Protein Electrophoresis Grade)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 100 g
DNase DNase free
Electroendosmosis (EEO) 0.1 max.
Gelation Temperature 34° to 45°C
Gel Strength 800g/cm2min.
Formula Weight 306.12
Grade DNase- and RNase-Free
Packaging Poly Bottle
Chemical Name or Material Agarose
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'