Learn More
Brady™ Aggressive Adhesive Multipurpose Nylon Labels
The B-499 nylon cloth label is made of tough and versatile, general-purpose label material. Print almost anywhere with a Brady™ portable label printer.
Brand:  Brady™ 170793
 View more versions of this product
Product Code. 17940680
  • 29.45 € / Each
Explore more special offers
Description
Description
  • Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike
Specifications
Specifications
UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Matte
Nylon Cloth
Non-sterile
BMP41, BMP51, M511
-40°F to +194°F
16 ft. x 0.375 in.
0.375 in.
M4C-375-499
Black on White
Continuous
Labels
Component and Equipment Labeling, Data and Telecommunications Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Wire and Cable Labeling
1 Tape/Cartridge
0.0065 in.
16 ft.
0.95 cm