Learn More
BTX™ AgilePulse™ In Vivo ID (Intra-Dermal) Generator Only
AgilePulse™ In Vivo ID (Intra-Dermal) Generator Only provides maximum efficiency DNA and RNA delivery to skin, muscle, and tumor tissues.
Brand:  BTX™ 470401N
Product Code. 15477390
  • 37115.00 € / Each
Explore more special offers
Description
Description
  • Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency
  • Simple user interface
  • Pulse Agile Advantage
  • Uniform, reliable electroporation
  • Simple, Fast Treatment
Specifications
Specifications
Vaccine Development, Intra-Dermal Gene Delivery, Intra-Tumor Gene Delivery
5 kHz to 1 Hz
Electroporation Generator
AgilePulse In Vivo ID (Intra-Dermal) Generator Only
15.7 in.
7.9 in.
Generator Only
0.200 to 1000 ms
0.050 to 10 ms
11.3 kg
32 cm
Electrode Connector Cable
Square Wave
In Vivo electroporation accessories
BTX Electroporation
50 to 1000 V
Touchscreen
40 cm
20 cm
50°F to 104°F
10°C to 40°C
25 lb.
12.6 in.
Non-sterile
Touch Screen Display, Footswitch
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Intended for Research use only. Not for use in diagnostic or clinical procedures.