Learn More
BTX™ AgilePulse PLUS Safety Stand
Adjustable Gap Safety Stand, specifically for AgilePulse PLUS Electroporators and BTX Flatpack chambers.
Brand:  BTX™ 470610N
Product Code. 16684862
  • 2890.00 € / Each
Explore more special offers
Description
Description
  • Connect to Agilepulse PLUS for safe delivery of high voltage electrical pulses to BTX Flatpack chambers
  • For optimum transfections up to volumes of 10 mL (Part Number 47-0206)
Specifications
Specifications
Large Volume Transfection
110/240 V
Touchscreen
Electrode Connector Cable
Square Wave
AgilePulse PLUS Electroporator
AgilePulse PLUS Safety Stand for Flatpack
Non-sterile
Digital User