missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents

AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.

Brand:  Applied Biosystems™ 4387391

 View more versions of this product

Product Code. 30057794

  • 1892.00 € / 1000 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Recommended for RNA pathogen amplification
  • Consistent amplification of RNA targets with high specificity and sensitivity
  • Optimized to work with your target-specific primers and probes
  • Contains ROX for quantitative fluorescent signal normalization

AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization. A Detection Enhancer is also provided as an optional reagent for amplification of templates with a high GC content or persistent secondary structure.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient for 1000 reactions of 25 μL, contains:
• 14 mL 2X RT-PCR Buffer
• 1100 μL 25X RT-PCR Enzyme Mix
• 25 mL Nuclease-free Water.
Primer-probe
High
AmpliTaq Gold DNA Polymerase
7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex
AgPath-ID™, Ambion™
1,000 reactions
Dry Ice
25X
1000 Reactions
14 mL
Works on all AB platforms, but is optimized for the 7500.
Tube
1-step RT-qPCR
Standard
ROX (Pre-mixed)
One-Step RT-PCR Reagent
ArrayScript™
1,000 Reactions
25 X
RNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Veterinary Use Only.