missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents with manual

AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.

Brand:  Applied Biosystems™ AM1005M

 View more versions of this product

Product Code. 17153279

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Recommended for RNA pathogen amplification
  • Consistent amplification of RNA targets with high specificity and sensitivity
  • Optimized to work with your target-specific primers and probes
  • Contains ROX for quantitative fluorescent signal normalization

AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sufficient for 100 reactions of 25 μL, contains:

• 1375 μL 2 x RT-PCR Buffer

• 110 μL 25 x RT-PCR Enzyme Mix

• 1.75 mL Nuclease-free Water.
Primer-probe
High
AmpliTaq Gold DNA Polymerase
7500 System
AgPath-ID™, Ambion™
100 Reactions
100 Reactions
25X
100 Reactions
Works on all AB platforms, but is optimized for the 7500.
Tube
1-step RT-qPCR
Standard
ROX (Pre-mixed)
One-Step RT-PCR Reagent
ArrayScript™
Pathogen Detection, Virus Detection
25 X
1375 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.