missing translation for 'onlineSavingsMsg'
Learn More

AGTR2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA3210

Product Code. 10008539

This item is not returnable. View return policy

Description

Description

PA3-210 detects AGTR2 in human samples. PA3-210 has successfully been used in Western blot, immunocytochemical, and immunohistochemical (paraffin) procedures. The PA3-210 immunogen is a synthetic peptide corresponding to the C-terminal amino acids C(349)RKSSSLREMETFVS(363) of AGTR2

Angiotensin II is a potent vasopressor hormone and a primary regulator of aldosterone secretion that acts through at least two types of receptors, AGTR1 and AGTR2. It is an important effector controlling blood pressure and volume in the cardiovascular system and plays a major role in the development of the mammalian kidney and urinary tract. Like AGTR1, AGTR2 is a seven transmembrane G protein-coupled receptor (GPCR), but AGTR2 does not demonstrate most of the classic features of GPCR signaling. AGTR2 is involved in a wide range of activities, including the induction of neurite outgrowth and the inhibition of cellular proliferation, in addition to the known function of mediation of vasoconstriction.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

AGTR2
Polyclonal
Unconjugated
Agtr2
AGTR2; AI316812; Angiotensin ii at2; angiotensin II receptor type 2; angiotensin II receptor, type 2; angiotensin II type 2 receptor; angiotensin II type-2 receptor; angiotensin receptor 2; AT<sub>2</sub>R; AT2; AT2 receptor; AT2R; AT2-R; ATGR2; AW107640; MRX88; type-2 angiotensin II receptor
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Conc. Not Determined
whole serum with 0.02% sodium azide
P50052
Agtr2
Synthetic peptide corresponding to the C-terminal amino acids C(349)RKSSSLREMETFVS(363) of AGTR2
RUO
186
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers