missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AIM V™ Medium, liquid (research grade)

AIM V™ Medium, liquid (research grade)
Brand: Gibco™ 12055091
This item is not returnable.
View return policy
Description
Contains L-glutamine, streptomycin sulfate at 50μg/mL, and gentamicin sulfate at 10μg/mL.
Specifications
Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes | |
Serum Free Medium (SFM) | |
Glutamine, Phenol Red, HSA | |
500 mL | |
Streptomycin Sulfate, Gentamicin | |
Store in refrigerator (2–8°C) and protect from light. |
Liquid | |
Sterile-filtered | |
AIM V™ | |
Room Temperature | |
Research Grade | |
Mammalian Cell Culture |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2953883 | Certificate of Analysis | 31/01/2025 | 12055083,12055091 |
2366186 | Certificate of Analysis | 29/01/2025 | 12055083,12055091 |
2390837 | Certificate of Analysis | 29/01/2025 | 12055083,12055091 |
2451531 | Certificate of Analysis | 29/01/2025 | 12055083,12055091 |
2451541 | Certificate of Analysis | 29/01/2025 | 12055083,12055091 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ AIM V™ Medium, liquid (research grade) > Quantity: 500 mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction