Learn More
Grant Instruments™ Air Cleaner Recirculator, 1 Lamp
Easy to use UV cleaner-recirculator ideal for air disinfection in hospitals, research laboratories and clinics. The UV air flow cleaner-recirculator consists of 1 germicidal UV lamp, a fan unit equipped with dust filters and a control unit confined in a flow through chamber.
Brand:  Grant Instruments™ UVR-M EU
Product Code. 17755149
  • 528.00 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Full user protection from direct UV radiation
  • Simple to operate
  • Convenient fixation on walls or mount on a movable tripod (optional)
  • Compact, sturdy construction for everyday use
  • Simple to operate with on/off switch
  • UV lamp operation indicator – indicator light turns off when the lamp needs replacing
Applications:
Hospitals (outpatient departments, operating rooms, emergency rooms, delivery rooms etc.), kindergartens, research   laboratories, veterinary clinics, schools, offices.
Specifications
Specifications
18 mW/cm2/sec, 14 m3/hour
1 Lamp
UVR-S
UV Cleaner-Recirculator