Learn More
Tuttnauer Air Compressor
The Air Compressor introduces pressurized air into the chamber for rapid cooling of liquids for models with a fast cooling system. Not needed if air pressure source is available on site.
Brand:  Tuttnauer PUM059-0023
 View more versions of this product
Product Code. 17869538
  • 1715.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Introduces pressurized air into the chamber for rapid cooling of liquids for models with a fast cooling system.
  • Not needed if air pressure source is available on site.
Specifications
Specifications
Air Compressor
50 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Air Compressor > Air Compressor; Frequency: 50 Hz