Learn More
Abeba™ Air-Cushion 32370 Shoes
Air cushion PU-sole with high slip resistance. Sanitized™ treated.
Brand:  Abeba™ 32370/46
 View more versions of this product
Product Code. 12683810
  • 84.90 € / 1 pair
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 O1 SRA
  • Microfiber
  • White
  • Slip resistance PU outsole sole
  • Integrated footbed
  • Sanitized treated
  • ESD dissipative according to DIN EN 61340
  • Instep area with elastic band
  • Decontaminable
Specifications
Specifications
CE EN ISO 20347:2012 O1 SRA
Microfiber
Slip-resistant
32370
White
46
Polyurethane