Learn More
Thermo Scientific™ Air Filter Kits
Protect your recirculating chillers from dusty environments with these Air Filter Kits.
Brand: Thermo Scientific™ 598000000008
This item is not returnable.
View return policy
Description
Protect your recirculating chillers from dusty environments with Thermo Scientific™ Air Filter Kits.
For use with Thermo Scientific recirculating chillers.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Air and fluid filter kit (100 micron) | |
ThermoFlex™ 3500 or 5000 Recirculating Chiller | |
Air and Fluid Filter |