missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Hostess™ 32 Toilet Tissue Bulk Pack
GREENER_CHOICE

Combines quality with practicality and economy. Hostess™ 32 Toilet Tissue Bulk Pack are soft, strong, and absorbent with a folded design to prevent ripping and reduce wastage.

Brand:  Kimberly-Clark Professional™ 8036

155.72 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12648865

  • 245.00 € / Pack of 32
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 2 ply sheets
  • 100% recycled folded toilet paper
  • Combines quality with practicality and economy
  • Fully biodegradable reducing the risk of blocked toilets
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Toilet Tissue
18.6 cm
11 cm
White
High Traffic Washrooms
2-ply
7.3 in.
4.3 in.
Paper
18.6 x 11 cm (7.3 x 4.3 in.)
32 Packs x 500 Sheets
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.