missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Air Valve V2.0

Air valves are part of the SCAT™ Safety Cap system used to protect solvents in supply bottles from external contamination. They provide safe pressure equalization with clean air for your solvent inlet.

Brand:  SCAT™ 317010

25.90 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 16403819

  • 32.80 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

An exchange indicator shows the elapsed service life, allowing for prompt replacement of the air valve after 6 months of use. Purified ventilation enables precise and reproducible analytical results in the HPLC/UHPLC process.

  • Blocks solvent vapors
  • Protects health and the environment
  • Exchange indicator displays next replacement time
  • Easy to change without tools
  • Fits any SCAT™ Safety Cap
Specifications

Specifications

0.25 in.
1 Pc.
1 Valve
V2.0
Red
Polyurethane
UNF 0.25 in., 28 G
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.