Learn More
Syngene™ Airclean 600 PCR Workstation
GREENER_CHOICE
Airclean 600 PCR Workstation with UVTect microprocessor controller and vertical laminar air flow.
Brand:  Syngene™ SYN-48PWLF-E
8849.25 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 16457260
  • 9315.00 € / Each
Explore more special offers
Description
Description
  • With UVTect microprocessor controller
  • Provides Class 100 vertical laminar air flow and timed uv light for sterilization
  • Includes HEPA filter, initial prefilter, fluorescent light, UV shelf with pipette holder, safety switch, and slip hatch
Specifications
Specifications
PCR Workstation
UV Controller
48 in.
UL, CSA, and CE approved
220 V
121.92 cm