Learn More
Syngene™ AirClean™ Microscope Enclosure with UVTect™ Microprocessor Controller
GREENER_CHOICE
The downflow AirClean™ Microscope Enclosure provides a clean work environment and a UV light for irradiation between processes. UVTect™ microprocessor controller provides one-touch operation of all enclosure features. This configuration does not provide operator protection.
Brand:  Syngene™ SYN-AC632TLFUVCMIC220-E
 View more versions of this product
Product Code. 17735904
  • 10370.00 € / Each
Explore more special offers
Description
Description
Standard Features
  • No ductwork required
  • No installation required - plugs directly into a standard 110V or 220V electrical outlet
  • Encloses equipment while retaining full operator access
  • UVTect™ Microprocessor Controller
  • ISO 5 vertical laminar flow air
  • UV and fluorescent lights
  • Seamless plastic design - no joints or gaps in construction
  • Lab event timer
  • HEPA filtration for capture of particulate to 0.3 microns in size
Unique Safety Features
  • Filter safety lockout to ensure correct filter replacement
  • Polycarbonate and polypropylene design reflects UV energy
  • Tested to UL 61010-1,CSA C22.2 No. 61010-1, and EN 61326-1
  • Brushless, sparkless blower is post-filter
  • Filter and electronics are located above the workspace, eliminating the chance of chemical spill
  • Real time monitoring of installed filtration bed
Specifications
Specifications
Microscope Enclosure
UVTect™ Microprocessor Controller
32 in.
813 mm