Learn More
Syngene™ AirClean™ Systems Dead Air Box with UVTect™ Controller
GREENER_CHOICE
The AirClean™ Systems Dead Air Box provides a circulation-free environment for the amplification and manipulation of DNA/RNA. Each dead air box is standard with the UVTect™ Microprocessor Controller and a 254 nm ultraviolet light source for irradiation between amplifications.
Brand:  Syngene™ SYN-AC632TDBC220-E
 View more versions of this product
Product Code. 17725904
  • 6715.00 € / Each
Explore more special offers
Description
Description
AirClean™ Systems Dead Air Boxes provide a circulation-free environment for the amplification and manipulation of DNA/RNA. Each dead air box is standard with the UVTect™ Microprocessor Controller and a 254 nm ultraviolet light source for irradiation between amplifications. A safety interlock switch disables the UV light when the sash is raised, preventing operator exposure. Seamless design and UV-reflective construction materials further ensure no harmful UV energy escapes the enclosure.
Standard Features
  • Polycarbonate and polypropylene design reflects UV energy
  • Shipped fully assembled
  • Overlapping polycarbonate sash to prevent UV light exposure
  • Sash safety switch automatically turns off UV light when sash is raised
  • Electronics are located above the workspace, eliminating the chance of a chemical spill entering these sensitive areas
  • Unique Safety Features
  • No ductwork required
  • No installation required - plugs directly into a standard 110V or 220V electrical outlet
  • UVTect™ Microprocessor Controller
  • UV and fluorescent lights
  • Seamless plastic design -no joints or gaps in construction
  • UV shelf with built-in pipettor holder
  • Lab event timer
  • Digital UV light timer 0.01 seconds to 59 minutes
  • Access port for electrical cords
Specifications
Specifications
Dead Air Box
UVTect™ Microprocessor Controller, 254 nm Ultraviolet Light Source
32 in.
Standard/Tall Polycarbonate UV Units
Tall Version
81 cm