missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Airless Wall-mounted Dispenser with Elbow

For use with Airless Bottle, without hand contact.

Brand:  Christeyns™ 460250

Product Code. 17849864

  • 20.35 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Conveniently store your Airless bottle using this wall-mount
Specifications

Specifications

Hand Sanitizer Dispenser
Bottles up to 6.25 in. dia.
Freestanding or Wall Mount
Hinged Lid with Front Access Tray
Chrome
Stainless Steel
1
1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.