missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Airtight Box with White Lid

Airtight Box with White Lid is ideal as delivery box and click and collect service.

Brand:  Gilac™ G602012

 View more versions of this product

Product Code. 17958901

  • 53.05 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Airtight container, by pack of 12, perfect as delivery box and click and collect service.
  • Keeps flavours in and odours out.
  • Clear box: contents are visible and recognizable.
  • Flexible, watertight lid.
  • PP clear container. LDPE lid
  • Available in 3 sizes, stackable.
  • Customisation available on request for a minimum of 500 units.
Specifications

Specifications

0.2 gal.
Airtight Lid
HDPE
Airtight Box
110 x 110 x 75 mm
4.3 in.
4.3 in.
3 in.
0.6 L
White
Rectangular
12 Pk.
110 mm
110 mm
75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.