missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 2mm, I.D.: 1.2mm, Thermo Scientific™

21.90 € - 45.00 €

Specifications

Chemical Name or Material Al-23 Insulating Tube
Name Note Both ends open (Thin Wall)
Form Tube
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11397508
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031488.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11317518
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031488.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11337518
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031488.DT
250 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11327518
View Documents Promotion Details
Thermo Scientific Alfa Aesar
031488.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Al-23 Insulating Tube
Both ends open (Thin Wall)
Tube
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications