missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Protective Tube, One End Closed, O.D.: 12mm, I.D.: 8mm, Wall.: 2mm, Thermo Scientific™

113.00 € - 610.00 €

Specifications

Chemical Name or Material Al-23 Protective Tube
Name Note One end closed
View More Specs

Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342688
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11322688
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.DU
270 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11372678
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.DY
375 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11302688
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.EE
530 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11392678
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.EK
740 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11332688
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.EQ
1030 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11382678
View Documents Promotion Details
Thermo Scientific Alfa Aesar
030048.FA
2030 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Al-23 Protective Tube
One end closed
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications