missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open (Thin Wall);OD (mm), 10;ID (mm), 9, Thermo Scientific Chemicals

36.68 € - 84.92 €

Specifications

Chemical Name or Material Al-23 Tube
Name Note Both ends open (Thin walls)
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338257
View Documents
Thermo Scientific Alfa Aesar
031709.DM
100 mm
36.68 €
100mm
Please to purchase this item. Need a web account? Register with us today!
11308257
View Documents
Thermo Scientific Alfa Aesar
031709.DR
200 mm
59.90 €
200mm
Please to purchase this item. Need a web account? Register with us today!
11318257
View Documents
Thermo Scientific Alfa Aesar
031709.DW
300 mm
84.92 €
300mm
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Al-23 Tube
Both ends open (Thin walls)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only