missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Tube, One End Closed, O.D.: 10mm, I.D.: 6mm, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 030069.DW
This item is not returnable.
View return policy
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Al-23 Tube | |
One end closed | |
300 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Al-23 Tube, One End Closed, O.D.: 10mm, I.D.: 6mm, Thermo Scientific™ > Size: 300mm; 1 Ea.
Spot an opportunity for improvement?Share a Content Correction