missing translation for 'onlineSavingsMsg'
Learn More
Albendazole sulfone, Thermo Scientific Chemicals Product Code.: 15463327

Albendazole sulfone, Thermo Scientific Chemicals

Product Code. 15463327
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15463327

Brand: Thermo Scientific Alfa Aesar H25917.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Albendazole are currently used for chemotherapeutic treatment of AE. Albendazole sulfone is used as scolicidal agents on hydatid cysts(in vitro).

Solubility
Soluble in DMSO, and methanol.

Notes
Stor
TRUSTED_SUSTAINABILITY

Specifications

CAS 75184-71-3
Melting Point 290°C to 292°C
Molecular Formula C12H15N3O4S
MDL Number MFCD00600775
Quantity 1 g
Synonym albendazole sulfone, unii-1uic88380g, methyl 5-propylsulfonyl 2-benzimidazolecarbamate, methyl 5-propylsulfonyl-1h-benzimidazol-2-yl carbamate, methyl-5-propylsulfonyl-1h-benzimidazol-2-yl carbamate, 5-propylsulfonyl-2-benzimidazolecarbamic acid methyl ester, methyl 6-propylsulfonyl-1h-benzo d imidazol-2-yl carbamate, methyl n-5-propane-1-sulfonyl-1h-1,3-benzodiazol-2-yl carbamate, 2-benzimidazolecarbamic acid, 5-propylsulfonyl-, methyl ester
Solubility Information Soluble in DMSO,and methanol.
InChI Key CLSJYOLYMZNKJB-UHFFFAOYSA-N
SMILES CCCS(=O)(=O)C1=CC=C2N=C(NC(=O)OC)NC2=C1
IUPAC Name methyl N-[6-(propane-1-sulfonyl)-1H-1,3-benzodiazol-2-yl]carbamate
Molecular Weight (g/mol) 297.33
PubChem CID 53174
ChEBI CHEBI:80620
Formula Weight 297.33
Chemical Name or Material Albendazole sulfone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10143556 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberH25917.03
Lot Number10143556 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH25917.03
compliance-icons
Product Identifier
  • Albendazole sulfone
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Albendazole sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Albendazole sulfone, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.