missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Albumine bovine serum, for biochemistry, additional reagent for IGSS, protease free

Product Code. 10013260 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
This item is not returnable. View return policy

Product Code. 10013260

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Albumin, bovine serum (BSA), CAS # 9048-46-8, is a small, stable biomolecule derived from bovine blood with wide applications in biochemistry, immunochemistry, pharmacokinetics, and cell culture studies. | CAS: 9048-46-8

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Albumin, bovine serum (BSA) is available as a protease-free, lyophilized powder that may be stably stored at 2-8°c
  • It is comprised of 583 amino acid residues, and has a molecular weight of 66.5 kDa

Applications

  • BSA is used as a protein standard in quantification assays, such as the Bradford protein assay
  • It is used to stabilize enzymes in restriction digestions in molecular biology experiments
  • It is a blocking agent in immunoblots thus increasing signal to noise ratios by quenching non-specific immunochemical reactions
  • BSA is a nutritive agent and used as a supplement in cell and microbial culture media
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9048-46-8
MDL Number MFCD00145743

Specifications

Color Beige-Yellow
Assay Percent Range 96.0% to 100.0% (protein [F:6.22]) on dry substance; 98% to 100% (Albumin, 7% solution [by cellulose acetate electrophoresis])
Packaging Glass bottle
Quantity 10 g
Physical Form Powder
Chemical Name or Material Albumine Bovine/Fraction V

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.