missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcohol, Anhydrous, Reagent, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
Description
- For Histological Use
Specifications
Specifications
Quantity | 500 mL |
Flash Point | 13.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 0.79kg |
Grade | BAKER ANALYZED™ ACS Reagent |
Chemical Name or Material | Alcohol, Anhydrous, Reagent |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
- H225-Highly flammable liquid and vapour.
- H371-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction