missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Alcoholmeter without Thermometer

For determining the percentage of alcohol by volume in liquids.

Brand:  Karl Hecht™ 46170001

Product Code. 10536162

  • 28.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Reference temperature of 20 °C
  • Range from 0 to 100: 1 vol%
  • 1% vol. divisions
  • Length of 260 mm
  • With paper scale and steel ballast
Specifications

Specifications

Alcohol Hydrometer
Paper scale
20°C
260 mm
Steel Metal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers