missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 488 Tyramide SuperBoost™ Kit, goat anti-rabbit IgG

Features poly-HRP-conjugated goat anti-rabbit IgG secondary antibody. Alexa Fluor™ 488 Tyramide SuperBoost™ Kit combine brightness of AlexaFluor dyes with high signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce s

Brand:  Invitrogen™ B40943

 View more versions of this product

Product Code. 15631902

  • 341.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).

Features:

  • High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
  • Simple protocol and detection using standard filters
  • Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
  • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments

Benefits:

  • Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
  • Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
  • Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
  • Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals

Shelf Life:

  • Stable for 6 months after receiving
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 kit sufficient for 50 microscope slides (18 x 18 mm), containing:

• Blocking buffer (1X), 22.5 mL
• Poly-HRP-conjugated goat anti-rabbit secondary antibody (1X), 22.5 mL
• Alexa Fluor tyramide reagent
• Hydrogen peroxide (stabilized 3% solution), 28.5 mL
• Reaction buffer (20X), 6 mL
• Reaction stop solution, 8 mg
• Dimethylsulfoxide (DMSO), 200 μL

Store at 2-8°C. When stored as directed, the product is stable for 6 months after receiving.
50 Slides
Approved for shipment at room temperature or on wet ice
Alexa Fluor 488
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3071395 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog NumberB40943
Lot Number2856099 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberB40943
Lot Number2747510 Certificate TypeCertificate of Analysis Date31/08/2023 Catalog NumberB40943
Lot Number2647831 Certificate TypeCertificate of Analysis Date14/04/2023 Catalog NumberB40943
Lot Number2497484 Certificate TypeCertificate of Analysis Date02/07/2022 Catalog NumberB40943
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Alexa Fluor™ 488 Tyramide SuperBoost™ Kit, goat anti-rabbit IgG > 50 slides

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.