missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Alexa Fluor™ 555 Tyramide SuperBoost™ Kit, goat anti-rabbit IgG

Features poly-HRP-conjugated goat anti-rabbit IgG secondary antibody. Alexa Fluor™ 555 Tyramide SuperBoost™ Kit combine brightness of AlexaFluor dyes with high signal amplification of a poly-HRP-mediated tyramide labeling reaction to produce s

Brand:  Invitrogen™ B40923

463.60 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15671892

  • 610.00 € / Pack of 150
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

SuperBoost™ tyramide signal amplification is most sensitive method for detection of low abundant targets in multiplexable fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC ), and in situ hybridization (ISH).
Features:
  • High sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
  • Simple protocol and detection using standard filters
  • Suitable for high-resolution multiplex images, co-label with DAPI, secondary antibodies, and other SuperBoost kits
  • Requires 10-100 times less primary antibody then standard ICC/IHC/ISH experiments
Benefits:
  • Enhancement of signal using Alexa Fluor tyramides: Utilize Alexa Fluor tyramides, which react with HRP to ultimately deposit bright and photostable Alexa Fluor dye on surrounding proteins and other similar molecules
  • Poly-HRP enhancement: Several HRP enzymes are conjugated with short polymers, enhancing signal by several fold over regular HRP systems
  • Reaction stop solution: Include an HRP stop solution to halt the HRP reaction
  • Reduction of background: Include blockers for elimination or reduction of endogenous peroxidase and fluorescent background signals
Shelf Life:
  • Stable for 6 months after receiving
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Alexa Fluor 555
1 kit sufficient for 150 microscope slides (18 x 18 mm), containing: Blocking buffer (1X), 22.5 mL
  • Poly-HRP-conjugated goat anti-rabbit secondary antibody (1X), 22.5 mL
  • Alexa Fluor tyramide reagent
  • Hydrogen pero
  • SuperBoost™
    150 slides
    Approved for shipment at Room Temperature or on Wet Ice
    Tyramide Kit
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use Only. Not for use in diagnostic procedures.