missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alexa Fluor™ 647-Biotin Control Peptide
Click to view available options
Quantity:
0.4 mL
Unit Size:
Each
Specifications
Specifications
| Product Line | Alexa Fluor |
| Color | Far-Red |
| Content And Storage | 100 nM Alexa Fluor 647-Biotin Control peptide in 10 mM HEPES, 137 mM NaCl, 2.7 mM KCl (HEPES buffered saline). Store at -20°C and protect from light. |
| Format | Frozen |
| Label or Dye | Alexa Fluor™ 647 |
| Label Type | Alexa Fluor Dyes |
| Quantity | 0.4 mL |
| Research Category | Drug Discovery, Immunology |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction