missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alexidine dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Selective inhibitor of protein tyrosine phosphatases localized to mitochondrion 1 (PTPMT1) (IC50 = 1.08 μM in vitro). Stimulates increased insulin secretion by β-cells in rat pancreatic islets. Displays antitcancer properties in FaDu cells. Also preserves functional hematopoietic stem cells ex vivo.
Specifications
Specifications
CAS | 1715-30-6 |
Molecular Formula | C26H58Cl2N10 |
MDL Number | MFCD00082369 |
Quantity | 50 mg |
Synonym | alexidine dihydrochloride, alexidine hydrochloride, dsstox_cid_25881, dsstox_rid_81197, dsstox_gsid_45881, 2,4,11,13-tetraazatetradecanediimidamide, n,n-bis 2-ethylhexyl-3,12-diimino-, dihydrochloride, 2,4,11,13-tetraazatetradecanediimidamide, n1,n14-bis 2-ethylhexyl-3,12-diimino-, hydrochloride 1:2, alexidine hcl, prestwick_42, n,n-bis 2-ethylhexyl-3,12-diimino-2,4,11,13-tetraazatetradecanediimidamide dihydrochloride |
InChI Key | BRJJFBHTDVWTCJ-UHFFFAOYNA-N |
SMILES | Cl.Cl.CCCCC(CC)CN=C(N)NC(N)=NCCCCCCN=C(N)NC(N)=NCC(CC)CCCC |
IUPAC Name | 1-{N''-[6-({amino[N''-(2-ethylhexyl)carbamimidamido]methylidene}amino)hexyl]carbamimidamido}-N'-(2-ethylhexyl)methanimidamide dihydrochloride |
Molecular Weight (g/mol) | 581.72 |
PubChem CID | 102678 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction