Learn More
Alfacalcidol-d6, MedChemExpress
MedChemExpress Alfacalcidol-d6 is the deuterium labeled Alfacalcidol. Alfacalcidol is a non-selective VDR activator.
Brand: Medchem Express HY-15332
This item is not returnable.
View return policy
Description
Alfacalcidol-d6 is the deuterium labeled Alfacalcidol. Alfacalcidol is a non-selective VDR activator.Specifications
Alfacalcidol-d6 | |
1 mg | |
Research | |
O[C@H](C/1=C)C[C@H](O)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])[C@]3(C)CCC\2 | |
406.67 | |
Research | |
Solid |
1641940-94-4 | |
White | |
C27H38D6O2 | |
406.67 | |
98.0% | |
Metabolism-sugar/lipid metabolism |