missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AlgiMatrix™ 6-Well, 3D Culture System, Flat-Bottom Microplate
Click to view available options
Quantity:
1 x 6 well plate
4 x 6 well plates
No. of Wells:
6
Unit Size:
Each
Description
AlgiMatrix™ 3D Culture System 6 well plate is a user-friendly, animal –free bioscaffold available for the development of high-fidelity cell culture models that are more predictive of disease states and drug responses. AlgiMatrix™ sponges are formulated using pharmaceutical-grade raw material from brown seaweed. Its pure, chemically defined, and nontoxic macro porous structure improved lot-to-lot consistency, superior cell loading and excellent nutrient delivery, without damage to cells. The AlgiMatrix™ sponge is biodegradable and is stable at room temperature.
The AlgiMatrix™ 3D Culture System is suitable for a broad range of procedures; including Multicellular tumor spheroid assays (MCTS), hepatocyte and cardiomyocyte organogenesis studies, co-culture studies, and high-throughput drug screening assays.
The AlgiMatrix™ 3D Culture System is suitable for a broad range of procedures; including Multicellular tumor spheroid assays (MCTS), hepatocyte and cardiomyocyte organogenesis studies, co-culture studies, and high-throughput drug screening assays.
Specifications
Specifications
| Lid | With Lid |
| Content And Storage | Contains 4 x 6 well AlgiMatrix plate and 50 ml firming buffer. Store at room temperature 15-30°C |
| Product Line | Algimatrix |
| Color | Clear |
| Product Type | Microplate |
| No. of Wells | 6 |
| Sterility | Sterile |
| Surface Treatment | AlgiMatrix 3D Culture System |
| Well Shape | Flat Bottom (F-well) |
| Purity or Quality Grade | Cell Culture Grade |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction