missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AlgiMatrix™ 96-Well, 3D Culture System, Flat-Bottom Microplate

Product Code. 10699353
Click to view available options
Quantity:
1 x 96 well plate
5 x 96 well plates
No. of Wells:
96
This item is not returnable. View return policy
This item is not returnable. View return policy

Color: Clear; Sterility: Sterile; Lid: With Lid

AlgiMatrix™ 3D Culture System is the first user-friendly non-animal-derived bioscaffold for three-dimensional (3D) cell culture. AlgiMatrix™ 3D Culture System creates a cell culture model system that more closely reflects normal cell morphology and behavior, to meet researchers' requirements for 3D cell culture in fields such as toxicology, drug development, cancer and stem cell research, development and morphogenesis, tissue and organ engineering, heart disease, diabetes, and Alzheimers disease.

AlgiMatrix™ 3D Culture System is supplied in a 96-well format that fits well into the workflow of both lower- and higher-throughput users. Scientists can inoculate cells directly into the sterile microtiter plates preloaded with lyophilized alginate sponge, with no pretreatments or preparations required. The combination of convenience, high capacity, and microscopic visualization makes AlgiMatrix™ 3D Culture System an ideal solution for broad basic research and drug discovery applications.

Applications:
AlgiMatrix™ 3D Culture System is suitable for many cell-based screening, drug discovery, and human cell therapy procedures, including the Multicellular Tumor Spheroid Assays (MCTS),1 hepatocyte 2-4 and cardiomyocyte organogenesis studies,5 co-culture studies,6 high-throughput drug screening assays,1 and embryonic stem cell 3D differentiation.7 Each lot is performance and quality tested in a cell-based toxicity assay.

Storage:
Room temperature.

References:
1. Kunz-Schughart, L., et al., (2004) J Biomolec Screening 9, 273.
2. Dvir-Ginzberg, M., et al. (2003) Tissue Engineering 9, 757.
3. Elkayam, T., et al. (2006) Tissue Engineering 12, 1357.
4. Kedem, A., et al. (2005) Tissue Engineering 11, 715.
5. Dar, A., et al. (2002) Biotechnology & Bioengineering 80, 305.
6. Kim, J. (2005) Seminars in Cancer Biology 15, 365.
7. Gerecht-Nir, S., et al. (2004) Biotechnology & Bioengineering 88, 313.
TRUSTED_SUSTAINABILITY

Specifications

Lid With Lid
Content And Storage Store at room temperature.
Packaging 1 Pack
Product Line Algimatrix
Color Clear
Product Type Microplate
No. of Wells 96
Sterility Sterile
Surface Treatment AlgiMatrix 3D Culture System
Well Shape Flat Bottom (F-well)
No. per Pack 1/Pk.
Purity or Quality Grade Cell Culture Grade
Quantity 1 x 96 well plate
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.