missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 6.0 μm

Alignflow™ Flow Cytometry Alignment Beads for Red Lasers, 6.0 μm

Brand:  Molecular Probes™ A16504

428.45 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.


Product Code. 15383966

  • 465.00 € / 3mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Molecular Probes™ AlignFlow™ flow cytometry alignment beads are reliable references for aligning, focusing, and calibrating flow cytometers.The 6.0μm Alignflow™ Flow Cytometry Alignment Beads for Red Lasers are fluorescent-dye infused microspheres that are excitable at 630–640 nm with emission in the range of 645–680 nm.

View information for all flow cytometry calibration and size reference beads.

AlignFlow™ beads provide:

  • Uniformity—very low FSC CV and intensity CV
  • Consistency in production, creating ideal reference standards
  • Confidence through reliable, optimal instrument performance
  • Compatibility—use with any instrument

    The AlignFlow™ beads are highly uniform with respect to size and fluorescence intensity, resulting in low FSC CV and intensity CV. The beads approximate the size, emission wavelength, and intensity of many biological samples and permit the calibration of the flow cytometer’s laser source, optics, and stream flow without wasting valuable and sensitive experimental material.

    Bead Labeling
    As with all of our flow cytometry standard microspheres, the AlignFlow™ beads are stained internally rather than on the surface. The dyes are therefore insulated from environmental interactions that could cause variable fluorescence output, resulting in excellent signal stability. These fluorescent polystyrene microspheres are supplied as suspensions packaged in dropper vials for convenient dispensation and are available in a choice of three colors (UV, blue, red) and two sizes (2.5μm, 6.0μm) to match your laser.

    Notes:
    The AlignFlow™ flow cytometry reference beads are provided in updated packaging that includes a Quick Reference Label that puts key information for product use at your fingertips.

    These beads replace previous AlignFlow™ flow cytometry reference bead products. This particular product replaces AlignFlow™ Plus Flow Cytometry Alignment Beads, 6.0μm, for 633 excitation (A7313).
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Contains 2 vials of Alignflow™ beads (1.5 mL per vial). Store at 2–8°C.
    6 μm
    Flow Cytometer
    3 mL
    Core
    Alignment Beads
    Fluorescence
    633 or 635
    Suspension
    Room Temperature
    AlignFlow™
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.