missing translation for 'onlineSavingsMsg'
Learn More

Aliquat∣r 336 TG

Product Code. 11443477
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11443477

Brand: Thermo Scientific Alfa Aesar A17247.AP

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63393-96-4 | C25H54ClN | 404.16 g/mol

Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aliquat 336 is widely utilized as a phase transfer catalyst for the catalytic oxidation of cyclohexene to 1,6-hexanedioic acid. It is involved in the total synthesis of Manzamine A. It serves as an anti-static agent for textile fabrics and carpeting. It finds application in waste water treatment to control foaming. It has a major application as a metal extraction reagent for cadmium, zinc, iron and rare earth metals. It is used as an effective catalyst for the preparation of methylphenyl ether from sodium phenolate with for methyl bromide.

Solubility
Miscible with benzene and chloroform.

Notes
Incompatible with strong oxidizing agents, acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63393-96-4
Molecular Formula C25H54ClN
MDL Number MFCD00011862
InChI Key XKBGEWXEAPTVCK-UHFFFAOYSA-M
Synonym methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
PubChem CID 21218
ChEBI CHEBI:75286
IUPAC Name methyl(trioctyl)azanium;chloride

Specifications

Quantity 500 mL
UN Number UN2810
SMILES [Cl-].CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol) 404.16
Formula Weight 404.17
Chemical Name or Material Aliquat 336
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Aliquat 336 TG
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Quaternary ammoniumcompounds, tri-C8- 10-alkylmethyl, chlorides, 1-Octanol, 1-Decanol, Amines, tri-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.