missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Alkaline Phosphatase Buffer-1 (5X)
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5
Specifications
Specifications
| Chemical Name or Material | Alkaline Phosphatase Buffer-1 (5X) |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Formulation | 200mM Tris, 200mM NaCl, and 40mM magnesium chloride at pH 9.5 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction