Learn More
Thermo Scientific™ Alkaline Saline Peptone Water (ISO) (Dehydrated)
Enrich and recover Vibrio species from food and water samples with this medium, while adhering to ISO/TS 21872-1&2:2007.
Brand:  Thermo Scientific™ CM1117B
Product Code. 11923362
  • 65.75 € / 500g
Estimated Shipment: 18-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Enrich and recover Vibrio species from food and water samples with Thermo Scientific™ Oxoid™ Alkaline Saline Peptone Water (ISO) (Dehydrated), while adhering to ISO/TS 21872-1&2:2007. This medium is selective by exploiting the ability of Vibrio species to tolerate alkaline conditions and high sodium chloride levels. The peptone content provides the conditions for growth and recovery of Vibrio species, supplying vitamins, minerals, and nitrogen in the form of amino acids.
Use Alkaline Saline Peptone Water (ISO) for selective recovery and enrichment for Vibrio species.
  • Creates strong alkaline and high sodium conditions to provide a selective environment for Vibrio species.
  • Adheres to the ISO/TS 21872-1&2 2007 standards.
  • Can be used as a non-selective enrichment broth for the cultivation of Aeromonas species.
Alkaline Saline Peptone Water (ISO) was first described by Shread, Donovan, and Lee1 to be used as a non-selective enrichment broth for the cultivation of Aeromonas species. Cruickshank reported that the raised pH of the medium could be used to effectively cultivate Vibrio species2.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10°C to 30°C
Powder
500 g
Alkaline Saline Peptone Water (ISO) (Dehydrated)
Pre-enrichment Medium