Learn More
Cytiva AlkPhos Direct™ Hybridization Buffer for Direct Labeling System
Fast and convenient system for all high-sensitivity applications using DNA, RNA, or oligonucleotide probes
Brand:  Cytiva RPN3688
743.85 EUR valid until 2024-12-31
Use promo code "21954" to get your promotional price.
Product Code. 10566875
  • 822.00 € / Each
Explore more special offers
Includes: Each system includes the following reagents, sufficient for 25 or 50 labeling reactions and detection of 5000cm2 membrane: labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer, blocking agent, CDP-Star™ or ECF detection reagents.
Description
Description
  • Hybridization stringency can be controlled by elevating temperatures up to 75°C (167°F), or by varying salt concentration
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • ECF™ detection for quantitation experiments
  • Thirty minutes from completion of hybridization to application of detection reagent: saves two to three hours over conventional indirect methods
  • Resulting probes can be used without purification
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fast, convenient for high-sensitivity applications with DNA, RNA, Oligo probes
Labeling reagent, crosslinker, control DNA, Reaction Buffer, hybridization Buffer, blocking agent, CDP-Star™ or ECF detection reagents
Amersham AlkPhos Direct Hybridization Buffer sufficient for 5000 cm2
For 5000 cm2 membrane
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1