Learn More
Brady™ All White Self-laminating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Self-laminating Vinyl is an ideal material for wire and cable identification. This material has good clarity, conformability and is self-extinguishing.
Brand:  Brady™ 173807
 View more versions of this product
Product Code. 17801369
  • 57.75 € / Pack of 250
Explore more special offers
Description
Description
  • Self-laminating vinyl is ideal for use in cable and wire labeling and general-purpose marking in labs
  • Durable, permanent acrylic adhesive has outstanding water and oil resistance
  • UL approved
  • Service temperature of -40°C to 70°C (-40°F to 158°F)
  • Made of B-427 vinyl
Specifications
Specifications
Not Autoclavable
M6-30-427-AW
None
Matte
Vinyl
Thermal Transfer
Non-sterile
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
250 Labels
-94°F to +158°F
0.004 in.
0.75 x 1.5 in.
1.5 in.
19.05 mm
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Permanent Acrylic
White
Die-cut
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
No
Rectangle
-70°C to +70°C
0.102 mm
0.75 in.
38.1 mm