missing translation for 'onlineSavingsMsg'
Learn More
Allantoin, 98%, Thermo Scientific Chemicals Product Code.: 10745741

Allantoin, 98%, Thermo Scientific Chemicals

Product Code. 10745741
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10745741

Brand: Thermo Scientific Acros 102841000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-59-6
Molecular Formula C4H6N4O3
Molecular Weight (g/mol) 158.12
MDL Number MFCD00005260
InChI Key POJWUDADGALRAB-UHFFFAOYSA-N
Synonym allantoin, 5-ureidohydantoin, glyoxyldiureide, 1-2,5-dioxoimidazolidin-4-yl urea, glyoxyldiureid, allantol, cordianine, sebical, alantan, avc/dienestrolcream
PubChem CID 204
ChEBI CHEBI:15676
IUPAC Name (2,5-dioxoimidazolidin-4-yl)urea
SMILES C1(C(=O)NC(=O)N1)NC(=O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 100 g
Beilstein 25, V,15, 338
Merck Index 15, 250
Solubility Information Solubility in water: 1g/190mL. Other solubilities: readily soluble in alkalies,1g/500mL alcohol,practically insoluble in ether
Formula Weight 158.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Allantoin, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462959 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog Number102841000,102845000
Lot NumberA0448181 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number102841000,102845000
Lot NumberA0435501 Certificate TypeCertificate of Analysis Date31/10/2021 Catalog Number102841000,102845000
Lot NumberA0397148 Certificate TypeCertificate of Analysis Date30/11/2020 Catalog Number102841000,102845000
Lot NumberA0421222 Certificate TypeCertificate of Analysis Date04/08/2020 Catalog Number102841000,102845000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allantoin, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.