missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente™ Allihn Condensers with Glass Olives

Allihn condensers for condensing vapours

Brand:  Lenz Laborglasintrumente™ 05223201

Product Code. 12643790

  • 69.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Allihn condensers acc. to DIN 12581 and in addition to DIN
  • With NS socket and NS cone
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications

Specifications

Cooling Surface - 100 cm²
6
NS 14/23
160 mm
1
DURAN™ Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers