missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloxan monohydrate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Color | Beige-Yellow to White |
Assay Percent Range | 98% |
Beilstein | 24, 500 |
Merck Index | 10, 276 |
Synonym | barbituric acid, 5,5-dihydroxy, 2,4,6 1h,3h,5h-pyrimidinetrione, 5,5-dihydroxy, unii-o2aap9f8b6, 5,5-dihydroxybarbituric acid, ccris 5957, mesoxalylurea monohydrate, 2,4,5,6 1h,3h-pyrimidinetetrone hydrate, 5,5-dihydroxyperhydropyrimidinetrione, 5,5-dihydroxy-2,4,6 1h,3h,5h-pyrimidinetrione, o2aap9f8b6 |
SMILES | C1(=O)C(C(=O)NC(=O)N1)(O)O |
IUPAC Name | 5,5-dihydroxy-1,3-diazinane-2,4,6-trione |
Molecular Weight (g/mol) | 160.085 |
Show More |
Safety and Handling
Product Identifier
- Alloxan monohydrate
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : 2,4,5,6(1H,3H)-Pyrimidinetetrone, monohydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'